Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179815.3 | NP_850146.1 | 11 - 12 | 2351 | 106 | |
GenBank mRNA | AB206553.1 | - | 11 - 12 | 2106 | 106 | |
AY062492.1 | - | 11 - 12 | 2114 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |