Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAH-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112484.5 | NP_188235.1 | 1 - 2 | 313 | 105 | |
GenBank mRNA | AJ245631.1 | - | 1 - 2 | 182 | 105 | |
AK226239.1 | - | 1 - 2 | 200 | 105 | ||
AY088822.1 | - | 1 - 2 | 196 | 105 | ||
BT028906.1 | - | 1 - 2 | 134 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |