Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338235.1 | NP_001325855.1 | 3 - 4 | 1013 | 149 | |
NM_112521.4 | NP_188271.1 | 3 - 4 | 1303 | 149 | ||
GenBank mRNA | AF332394.2 | - | 3 - 4 | 708 | 149 | |
AF332401.1 | - | 3 - 4 | 708 | 149 | ||
AF386948.1 | - | 3 - 4 | 844 | 149 | ||
AY072502.1 | - | 3 - 4 | 708 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |