Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUC-L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338347.1 | NP_001325664.1 | 12 - 13 | 1255 | 139 | |
NM_001338348.1 | NP_001325663.1 | 13 - 14 | 1288 | 139 | ||
NM_112747.3 | NP_188491.1 | 14 - 15 | 1320 | 139 | ||
GenBank mRNA | DQ446672.1 | - | 14 - 15 | 1311 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |