Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112791.6 | NP_188535.4 | 22 - 23 | 4318 | 114 | |
GenBank mRNA | DQ399530.1 | - | 21 - 22 | 4118 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |