Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 13 - 14 | 1799 | 95 | |
NM_001338387.1 | NP_001325757.1 | 13 - 14 | 1780 | 95 | ||
NM_112808.4 | NP_188552.3 | 13 - 14 | 1780 | 95 | ||
GenBank mRNA | AB250666.1 | - | 13 - 14 | 1755 | 95 | |
DQ912973.1 | - | 13 - 14 | 1693 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |