Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084722.1 | NP_001078191.1 | 11 - 12 | 1355 | 115 | |
NM_112893.4 | NP_188637.2 | 12 - 13 | 1421 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |