Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342328.1 | NP_001328042.1 | 1 - 2 | 480 | 80 | |
NM_119684.4 | NP_567977.2 | 1 - 2 | 480 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |