Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204003.1 | NP_001190932.1 | 1 - 2 | 350 | 66 | |
NM_119774.3 | NP_195331.2 | 1 - 2 | 429 | 66 | ||
GenBank mRNA | DQ446901.1 | - | 1 - 2 | 318 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |