Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119857.3 | NP_568010.1 | 6 - 7 | 1153 | 73 | |
GenBank mRNA | AF319540.1 | - | 6 - 7 | 1091 | 73 | |
AK229267.1 | - | 6 - 7 | 1109 | 73 | ||
BT026462.1 | - | 6 - 7 | 1069 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |