Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202751.1 | NP_001189680.1 | 8 - 9 | 680 | 143 | |
NM_179908.3 | NP_850239.1 | 7 - 8 | 769 | 143 | ||
GenBank mRNA | AJ312202.1 | - | 7 - 8 | 769 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |