Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035850.3 | NP_001030927.1 | 6 - 7 | 603 | 112 | |
NM_001331269.1 | NP_001322867.1 | 7 - 8 | 516 | 112 | ||
NM_001331270.1 | NP_001322868.1 | 7 - 8 | 486 | 112 | ||
NM_100019.2 | NP_563627.1 | 7 - 8 | 516 | 112 | ||
GenBank mRNA | AB081500.1 | - | 6 - 7 | 328 | 112 | |
AF465800.1 | - | 6 - 7 | 319 | 112 | ||
AK175840.1 | - | 6 - 7 | 375 | 112 | ||
BT024771.1 | - | 6 - 7 | 319 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |