Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331454.1 | NP_001322971.1 | 1 - 2 | 313 | 127 | |
NM_001331455.1 | NP_001322970.1 | 1 - 2 | 313 | 127 | ||
NM_100279.5 | NP_171894.1 | 1 - 2 | 313 | 127 | ||
GenBank mRNA | AY044049.1 | - | 1 - 2 | 311 | 127 | |
BT029214.1 | - | 1 - 2 | 76 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |