Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331602.1 | NP_001322445.1 | 1 - 2 | 300 | 94 | |
NM_001331603.1 | NP_001322446.1 | 1 - 2 | 303 | 94 | ||
NM_100475.4 | NP_563754.1 | 1 - 2 | 313 | 94 | ||
GenBank mRNA | AF360321.1 | - | 1 - 2 | 289 | 94 | |
AY113890.1 | - | 1 - 2 | 255 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |