Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT71C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100598.4 | NP_172204.1 | 1 | 1716 | 160 | |
GenBank mRNA | AY065190.1 | - | 1 | 1472 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |