Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125053.1 | NP_001118525.1 | 6 - 7 | 1039 | 130 | |
NM_001337088.1 | NP_001324585.1 | 7 - 8 | 904 | 130 | ||
NM_001337089.1 | NP_001324587.1 | 7 - 8 | 904 | 130 | ||
NM_001337090.1 | NP_001324586.1 | 2 - 3 | 518 | 130 | ||
NM_130011.3 | NP_181976.1 | 7 - 8 | 843 | 130 | ||
GenBank mRNA | AY074629.1 | - | 7 - 8 | 814 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |