Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331741.1 | NP_001318950.1 | 8 - 9 | 1301 | 74 | |
NM_100665.3 | NP_172269.2 | 8 - 9 | 1179 | 74 | ||
GenBank mRNA | AK230326.1 | - | 8 - 9 | 1146 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |