Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331853.1 | NP_001320712.1 | 2 - 3 | 433 | 105 | |
NM_001331854.1 | NP_001320713.1 | 5 - 6 | 631 | 105 | ||
NM_100854.5 | NP_172452.3 | 6 - 7 | 550 | 105 | ||
GenBank mRNA | AK221696.1 | - | 6 - 7 | 515 | 105 | |
AK226991.1 | - | 6 - 7 | 552 | 105 | ||
BT029759.1 | - | 2 - 3 | 264 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |