Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331926.1 | NP_001318975.1 | 18 - 19 | 2638 | 71 | |
NM_001331927.1 | NP_001320611.1 | 17 - 18 | 2487 | 71 | ||
NM_100952.4 | NP_563877.1 | 19 - 20 | 2472 | 71 | ||
GenBank mRNA | AF312027.1 | - | 16 - 17 | 2178 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |