Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331946.1 | NP_001320672.1 | 1 - 2 | 384 | 70 | |
GenBank mRNA | AK221596.1 | - | 2 - 3 | 272 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |