Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203644.1 | NP_001190573.1 | 15 - 16 | 2342 | 64 | |
NM_125397.3 | NP_200812.2 | 15 - 16 | 2187 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |