Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125447.2 | NP_568922.1 | 4 - 5 | 684 | 126 | |
GenBank mRNA | AK117313.1 | - | 4 - 5 | 684 | 126 | |
AY087902.1 | - | 4 - 5 | 592 | 126 | ||
BT005266.1 | - | 4 - 5 | 529 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |