Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335968.1 | NP_001325298.1 | 1 - 2 | 201 | 68 | |
NM_128058.4 | NP_180072.1 | 1 - 2 | 201 | 68 | ||
GenBank mRNA | AK227987.1 | - | 1 - 2 | 113 | 68 | |
BT004611.1 | - | 1 - 2 | 51 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |