Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 14 - 15 | 1847 | 103 | |
GenBank mRNA | AY035110.1 | - | 14 - 15 | 1156 | 103 | |
FJ708701.1 | - | 14 - 15 | 1080 | 103 | ||
U47029.1 | - | 14 - 15 | 1130 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |