Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 1 - 2 | 844 | 127 | |
GenBank mRNA | AY035110.1 | - | 1 - 2 | 153 | 127 | |
FJ708701.1 | - | 1 - 2 | 77 | 127 | ||
U47029.1 | - | 1 - 2 | 127 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |