Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036347.2 | NP_001031424.1 | 9 - 10 | 960 | 79 | |
NM_001036348.2 | NP_001031425.1 | 10 - 11 | 1256 | 79 | ||
NM_001336077.1 | NP_001325357.1 | 10 - 11 | 793 | 79 | ||
NM_128216.3 | NP_565626.1 | 10 - 11 | 1221 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |