Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128498.5 | NP_180505.1 | 1 - 2 | 453 | 77 | |
GenBank mRNA | AF288187.1 | - | 1 - 2 | 321 | 77 | |
AK229012.1 | - | 1 - 2 | 370 | 77 | ||
BT024843.1 | - | 1 - 2 | 321 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |