Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338499.1 | NP_001319603.1 | 10 - 11 | 1670 | 62 | |
NM_001338500.1 | NP_001327317.1 | 10 - 11 | 1566 | 62 | ||
NM_202618.3 | NP_974347.1 | 10 - 11 | 1501 | 62 | ||
GenBank mRNA | AK176132.1 | - | 10 - 11 | 1331 | 62 | |
AY383545.1 | - | 10 - 11 | 1361 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |