Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKP-ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113196.3 | NP_566720.1 | 2 - 3 | 885 | 81 | |
GenBank mRNA | AY056793.1 | - | 2 - 3 | 809 | 81 | |
AY058084.1 | - | 2 - 3 | 836 | 81 | ||
BT002329.1 | - | 2 - 3 | 701 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |