Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338695.1 | NP_001319633.1 | 2 - 3 | 641 | 69 | |
NM_113370.3 | NP_189102.3 | 2 - 3 | 673 | 69 | ||
GenBank mRNA | BT015406.1 | - | 2 - 3 | 611 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |