Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP86C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113519.2 | NP_189243.1 | 1 | 1534 | 111 |
Gene Symbol: AT3G26130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338789.1 | NP_001327393.1 | 3 | 2081 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |