Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI-4KBETA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345618.1 | NP_001330874.1 | 1 - 2 | 2108 | 110 | |
NM_125803.4 | NP_201212.1 | 1 - 2 | 2140 | 110 | ||
GenBank mRNA | AF462854.1 | - | 1 - 2 | 2108 | 110 | |
AY139795.1 | - | 1 - 2 | 1564 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |