Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101026.3 | NP_172620.1 | 2 - 3 | 325 | 102 | |
GenBank mRNA | AF144390.1 | - | 2 - 3 | 210 | 102 | |
BT024568.1 | - | 2 - 3 | 203 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |