Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331996.1 | NP_001323168.1 | 1 | 416 | 102 | |
NM_101030.4 | NP_172623.1 | 1 | 303 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |