Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332096.1 | NP_001318998.1 | 2 - 3 | 436 | 105 | |
NM_101255.2 | NP_172842.1 | 1 - 2 | 505 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |