Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084068.3 | NP_001077537.1 | 7 - 8 | 1360 | 129 | |
NM_101338.4 | NP_172922.2 | 6 - 7 | 1443 | 129 | ||
GenBank mRNA | AY120768.1 | - | 6 - 7 | 1378 | 129 | |
BT008373.1 | - | 6 - 7 | 1080 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |