Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106472.1 | NP_565174.1 | 1 - 2 | 61 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |