Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 13 - 14 | 3799 | 74 | |
NM_001334833.1 | NP_001322931.1 | 12 - 13 | 3758 | 74 | ||
NM_001334834.1 | NP_001322932.1 | 12 - 13 | 3725 | 74 | ||
NM_106505.5 | NP_177979.1 | 12 - 13 | 1989 | 74 | ||
GenBank mRNA | KJ138926.1 | - | 11 - 12 | 1783 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |