Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106630.3 | NP_565222.1 | 3 - 4 | 569 | 59 | |
NM_202454.2 | NP_974183.1 | 3 - 4 | 573 | 59 | ||
GenBank mRNA | AY064063.1 | - | 3 - 4 | 570 | 59 | |
AY096373.1 | - | 3 - 4 | 369 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |