Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111840.3 | NP_187616.1 | 5 - 6 | 1620 | 109 | |
GenBank mRNA | AF096281.1 | - | 5 - 6 | 1313 | 109 | |
AY065037.1 | - | 5 - 6 | 1332 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |