Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337871.1 | NP_001327644.1 | 5 - 6 | 803 | 92 | |
NM_111873.4 | NP_566372.1 | 5 - 6 | 883 | 92 | ||
GenBank mRNA | AK118541.1 | - | 5 - 6 | 707 | 92 | |
AY059763.1 | - | 5 - 6 | 707 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |