Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338928.1 | NP_001319661.1 | 4 - 5 | 722 | 73 | |
NM_001338929.1 | NP_001325749.1 | 4 - 5 | 722 | 73 | ||
NM_001338930.1 | NP_001325748.1 | 3 - 4 | 726 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |