Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338993.1 | NP_001326021.1 | 4 - 5 | 445 | 115 | |
NM_113818.4 | NP_189539.1 | 4 - 5 | 538 | 115 | ||
GenBank mRNA | AK118926.1 | - | 4 - 5 | 443 | 115 | |
BT005690.1 | - | 4 - 5 | 304 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |