Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035717.2 | NP_001030794.1 | 2 - 3 | 628 | 77 | |
NM_001084753.2 | NP_001078222.1 | 2 - 3 | 614 | 77 | ||
NM_113878.5 | NP_189598.1 | 1 - 2 | 876 | 77 | ||
GenBank mRNA | AF385741.1 | - | 1 - 2 | 719 | 77 | |
AK317411.1 | - | 2 - 3 | 483 | 77 | ||
AY133676.1 | - | 1 - 2 | 388 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |