Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160745.1 | NP_001154217.1 | 7 - 8 | 619 | 111 | |
NM_001340630.1 | NP_001319888.1 | 4 - 5 | 750 | 111 | ||
NM_001340631.1 | NP_001328831.1 | 4 - 5 | 762 | 111 | ||
NM_117040.4 | NP_192710.1 | 5 - 6 | 641 | 111 | ||
NM_179016.1 | NP_849347.1 | 3 - 4 | 409 | 111 | ||
GenBank mRNA | AB071851.1 | - | 5 - 6 | 402 | 111 | |
AK229171.1 | - | 5 - 6 | 532 | 111 | ||
BT029504.1 | - | 5 - 6 | 402 | 111 | ||
BT030039.1 | - | 3 - 4 | 300 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |