Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117086.4 | NP_192756.2 | 1 - 2 | 204 | 82 | |
GenBank mRNA | BT015348.1 | - | 1 - 2 | 107 | 82 | |
BT020454.1 | - | 1 - 2 | 72 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |