Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCS52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117262.3 | NP_192929.2 | 7 - 8 | 1477 | 99 | |
GenBank mRNA | AY063875.1 | - | 7 - 8 | 1432 | 99 | |
AY091235.1 | - | 7 - 8 | 1307 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |