Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117266.4 | NP_192933.2 | 1 - 2 | 349 | 133 | |
GenBank mRNA | AK117721.1 | - | 1 - 2 | 219 | 133 | |
BT005305.1 | - | 1 - 2 | 170 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |