Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118996.5 | NP_567812.1 | 2 - 3 | 680 | 109 | |
GenBank mRNA | AY099715.1 | - | 2 - 3 | 420 | 109 | |
AY943845.1 | - | 2 - 3 | 131 | 109 | ||
BT000286.1 | - | 2 - 3 | 131 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |