Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102124.3 | NP_564180.1 | 10 - 11 | 3257 | 68 | |
GenBank mRNA | AF105064.1 | - | 9 - 10 | 3029 | 68 | |
AJ133786.1 | - | 9 - 10 | 2819 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |